Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Macc1
Entrez Gene ID
100753453
Gene Name
metastasis associated in colon cancer 1
Gene Aliases
I79_020678
Chromosome Location
Chr.Un|NW_003615314: 242943 - 295580 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512017.2 XP_003512065.1 2 - 3 176 82

View Full Product Details