Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ska2
Entrez Gene ID
100755178
Gene Name
spindle and kinetochore associated complex subunit 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614004: 753329 - 770926 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503803.2 XP_003503851.2 3 - 4 356 129

View Full Product Details