Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1ql3
Entrez Gene ID
100767576
Gene Name
complement component 1, q subcomponent-like 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614115: 856721 - 865099 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505038.2 XP_003505086.2 1 - 2 125 125

View Full Product Details