Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kcnj14
Entrez Gene ID
100775100
Gene Name
potassium inwardly-rectifying channel, subfamily J, member 14
Gene Aliases
I79_020928
Chromosome Location
Chr.Un|NW_003615387: 53659 - 59355 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652684.1 XP_007650874.1 1 - 1 424 128

View Full Product Details