Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1qtnf5
Entrez Gene ID
100760072
Gene Name
C1q and tumor necrosis factor related protein 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616562: 98497 - 100576 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654530.1 XP_007652720.1 1 - 1 114 82
XM_007654531.1 XP_007652721.1 1 - 1 114 82

View Full Product Details