Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB16
Entrez Gene ID
102178465
Gene Name
zinc finger and BTB domain containing 16
Gene Aliases
PLZF
Chromosome Location
Chr.15: 57556179 - 57759739 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314273.1 NP_001301202.1 5 - 6 1871 69
XM_018058854.1 XP_017914343.1 6 - 7 2376 69
XM_018058856.1 XP_017914345.1 6 - 7 2095 69
XM_018058857.1 XP_017914346.1 5 - 6 1927 69
GenBank mRNA JT743172.1 - 5 - 6 2018 69
JX047313.1 - 5 - 6 1871 69

View Full Product Details