Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
DLX5
Entrez Gene ID
102184778
Gene Name
distal-less homeobox 5
Gene Aliases
-
Chromosome Location
Chr.4: 106264729 - 106269160 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047390.1 XP_017902879.1 2 - 3 813 86

View Full Product Details