Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNTTIP2
Entrez Gene ID
102186650
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 70920294 - 70934289 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678082.3 XP_005678139.2 6 - 7 2552 81

View Full Product Details