Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
CNFN
Entrez Gene ID
106503129
Gene Name
cornifelin
Gene Aliases
-
Chromosome Location
Chr.18: 51805738 - 51808057 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062505.1 XP_017917994.1 2 - 3 252 76
GenBank mRNA JT746069.1 - 2 - 3 240 76
KA326289.1 - 3 - 4 495 76

View Full Product Details