Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR37
Entrez Gene ID
102190018
Gene Name
G protein-coupled receptor 37
Gene Aliases
-
Chromosome Location
Chr.4: 31131109 - 31149455 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679413.3 XP_005679470.3 1 - 2 1864 64
GenBank mRNA JO631830.2 - 1 - 2 1112 64

View Full Product Details