Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEYL
Entrez Gene ID
102191579
Gene Name
hes related family bHLH transcription factor with YRPW motif like
Gene Aliases
-
Chromosome Location
Chr.3: 14351374 - 14367153 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042164.1 XP_017897653.1 4 - 5 926 74
XM_018042166.1 XP_017897655.1 4 - 5 364 74
XM_018042168.1 XP_017897657.1 4 - 5 310 74

View Full Product Details