Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRSS54
Entrez Gene ID
102174866
Gene Name
serine protease 54
Gene Aliases
-
Chromosome Location
Chr.18: 27461088 - 27476224 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692041.1 XP_005692098.1 5 - 6 926 77

View Full Product Details