Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXW12
Entrez Gene ID
102173299
Gene Name
F-box and WD repeat domain containing 12
Gene Aliases
-
Chromosome Location
Chr.8: 89819767 - 89855430 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684240.3 XP_005684297.1 2 - 3 174 60

View Full Product Details