Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NHS
Entrez Gene ID
102181284
Gene Name
NHS actin remodeling regulator
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 52707593 - 52983542 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043756.1 XP_017899245.1 8 - 9 4723 95
XM_018043757.1 XP_017899246.1 7 - 8 4585 95
GenBank mRNA JO430657.2 - 1 - 2 823 95

View Full Product Details