Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNB2
Entrez Gene ID
102179250
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Gene Aliases
-
Chromosome Location
Chr.3: 104132423 - 104144025 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046194.1 XP_017901683.1 2 - 3 459 77
GenBank mRNA JO645704.2 - 2 - 3 247 77

View Full Product Details