Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHX4
Entrez Gene ID
102189505
Gene Name
LIM homeobox 4
Gene Aliases
-
Chromosome Location
Chr.16: 59998545 - 60044375 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690939.2 XP_005690996.1 2 - 3 469 65

View Full Product Details