Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFAP3
Entrez Gene ID
102172537
Gene Name
microfibril associated protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 45031200 - 45048035 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013965713.2 XP_013821167.1 1 - 2 474 76
GenBank mRNA JO436592.2 - 1 - 2 805 76

View Full Product Details