Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPY19L2
Entrez Gene ID
102191398
Gene Name
dpy-19 like 2
Gene Aliases
-
Chromosome Location
Chr.4: 58249977 - 58343819 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679195.2 XP_005679252.1 3 - 4 647 77

View Full Product Details