Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGSN
Entrez Gene ID
102186060
Gene Name
lengsin, lens protein with glutamine synthetase domain
Gene Aliases
-
Chromosome Location
Chr.2: 136330970 - 136396341 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675803.3 XP_005675860.2 4 - 5 528 70

View Full Product Details