Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGF20
Entrez Gene ID
102171403
Gene Name
fibroblast growth factor 20
Gene Aliases
-
Chromosome Location
Chr.27: 25329576 - 25339635 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698852.2 XP_005698909.2 1 - 2 894 94

View Full Product Details