Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTC
Entrez Gene ID
100860880
Gene Name
ornithine carbamoyltransferase
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 32417561 - 32460354 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285729.1 NP_001272658.1 - 726 93
GenBank mRNA GQ148781.1 - - 726 93
JO419477.2 - - 1317 93

View Full Product Details