Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRSP-2
Entrez Gene ID
100860822
Gene Name
calcitonin receptor-stimulating peptide-2
Gene Aliases
CRSP2
Chromosome Location
Chr.15: 44708222 - 44713757 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285687.1 NP_001272616.1 4 - 5 435 89
XM_013969432.1 XP_013824886.1 4 - 5 516 88
GenBank mRNA AB364647.1 - 4 - 5 435 89

View Full Product Details