Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSG4
Entrez Gene ID
102182843
Gene Name
desmoglein 4
Gene Aliases
-
Chromosome Location
Chr.24: 26034208 - 26074385 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314293.1 NP_001301222.1 2 - 3 81 63
GenBank mRNA KM369171.1 - 2 - 3 81 63

View Full Product Details