Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RARRES2
Entrez Gene ID
102190100
Gene Name
retinoic acid receptor responder 2
Gene Aliases
chemerin
Chromosome Location
Chr.4: 7917257 - 7921039 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001312693.1 NP_001299622.1 1 - 2 192 85
XM_013963442.2 XP_013818896.1 2 - 3 310 85
XM_018046618.1 XP_017902107.1 2 - 3 325 85
XM_018046619.1 XP_017902108.1 2 - 3 224 85
XM_018046620.1 XP_017902109.1 2 - 3 316 85
XM_018046621.1 XP_017902110.1 2 - 3 324 85
XM_018046622.1 XP_017902111.1 2 - 3 324 85
XM_018046623.1 XP_017902112.1 2 - 3 224 85
XM_018046625.1 XP_017902114.1 2 - 3 324 85
XM_018046626.1 XP_017902115.1 2 - 3 324 85
GenBank mRNA JO443485.2 - 4 - 5 502 85
KT313029.1 - 1 - 2 192 85

View Full Product Details