Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB7A
Entrez Gene ID
102168869
Gene Name
RAB7A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.22: 59104355 - 59149577 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286091.1 NP_001273020.1 1 - 2 64 78
XM_018066621.1 XP_017922110.1 2 - 3 257 78
GenBank mRNA JO581437.1 - 4 - 5 1845 78
JQ417405.1 - 1 - 2 64 78

View Full Product Details