Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAMP
Entrez Gene ID
102170629
Gene Name
hepcidin antimicrobial peptide
Gene Aliases
-
Chromosome Location
Chr.18: 46821826 - 46823365 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013971234.2 XP_013826688.1 2 - 3 299 98

View Full Product Details