Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102180271
Entrez Gene ID
102180271
Gene Name
antigen-presenting glycoprotein CD1d
Gene Aliases
CD1D
Chromosome Location
Chr.3: 107889082 - 107892937 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677291.3 XP_005677348.2 2 - 3 934 64

View Full Product Details