Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPS27L
Entrez Gene ID
102172630
Gene Name
ribosomal protein S27 like
Gene Aliases
-
Chromosome Location
Chr.10: 56223042 - 56226521 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005685681.3 XP_005685738.1 2 - 3 252 78
GenBank mRNA KA335117.1 - 2 - 3 550 78

View Full Product Details