Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102180468
Entrez Gene ID
102180468
Gene Name
taste receptor type 2 member 10
Gene Aliases
T2R10
Chromosome Location
Chr.5: 97395587 - 97396516 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680832.1 XP_005680889.1 1 - 1 418 82

View Full Product Details