Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRK1
Entrez Gene ID
102176894
Gene Name
G protein-coupled receptor kinase 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189962: 305185 - 318771 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044994.1 XP_017900483.1 6 - 7 1630 116

View Full Product Details