Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUSD5
Entrez Gene ID
102172105
Gene Name
sushi domain containing 5
Gene Aliases
-
Chromosome Location
Chr.22: 7392064 - 7438390 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038257.1 XP_017893746.1 2 - 3 278 97

View Full Product Details