Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINK4
Entrez Gene ID
102189035
Gene Name
serine peptidase inhibitor, Kazal type 4
Gene Aliases
-
Chromosome Location
Chr.8: 74799649 - 74807171 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684054.3 XP_005684111.1 1 - 2 124 71

View Full Product Details