Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C15H11orf1
Entrez Gene ID
102171444
Gene Name
chromosome 15 C11orf1 homolog
Gene Aliases
-
Chromosome Location
Chr.15: 60140584 - 60144638 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689435.2 XP_005689492.1 2 - 3 627 93

View Full Product Details