Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMCD1
Entrez Gene ID
102190272
Gene Name
LIM and cysteine rich domains 1
Gene Aliases
-
Chromosome Location
Chr.22: 17740027 - 17796767 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695648.2 XP_005695705.1 5 - 6 1058 84
GenBank mRNA JO369355.2 - 5 - 6 1065 84
JO513736.2 - 1 - 2 672 84

View Full Product Details