Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL26
Entrez Gene ID
102187446
Gene Name
methyltransferase like 26
Gene Aliases
-
Chromosome Location
Chr.25: 482229 - 484666 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697339.3 XP_005697396.1 1 - 2 640 88
GenBank mRNA JO553607.2 - 5 - 6 567 88

View Full Product Details