Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINA4
Entrez Gene ID
102173613
Gene Name
serpin family A member 4
Gene Aliases
-
Chromosome Location
Chr.21: 58646742 - 58654146 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695357.3 XP_005695414.2 3 - 4 1085 62

View Full Product Details