Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ZBTB47
Entrez Gene ID
102179893
Gene Name
zinc finger and BTB domain containing 47
Gene Aliases
-
Chromosome Location
Chr.22: 14632641 - 14644905 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066825.1 XP_017922314.1 3 - 4 1864 54
XM_018066826.1 XP_017922315.1 3 - 4 1743 54

View Full Product Details