Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAC8
Entrez Gene ID
102178994
Gene Name
placenta associated 8
Gene Aliases
-
Chromosome Location
Chr.6: 98339602 - 98373344 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681847.3 XP_005681904.1 3 - 4 381 69
GenBank mRNA JO580108.1 - 2 - 3 475 69

View Full Product Details