Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLCO6A1
Entrez Gene ID
102178541
Gene Name
solute carrier organic anion transporter family member 6A1
Gene Aliases
-
Chromosome Location
Chr.7: 9124263 - 9219243 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051637.1 XP_017907126.1 8 - 9 1278 109

View Full Product Details