Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS14
Entrez Gene ID
102181976
Gene Name
mitochondrial ribosomal protein S14
Gene Aliases
-
Chromosome Location
Chr.16: 54751606 - 54773939 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690908.3 XP_005690965.3 2 - 3 294 85
GenBank mRNA JO577054.1 - 2 - 3 294 85

View Full Product Details