Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
NT5C
Entrez Gene ID
102172480
Gene Name
5', 3'-nucleotidase, cytosolic
Gene Aliases
-
Chromosome Location
Chr.19: 55540485 - 55542300 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063836.1 XP_017919325.1 3 - 4 458 97
GenBank mRNA KA313652.1 - 2 - 3 455 97

View Full Product Details