Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRAF2
Entrez Gene ID
102169086
Gene Name
PRA1 domain family member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017190143: 52346 - 55440 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700757.3 XP_005700814.3 1 - 2 593 77
GenBank mRNA JO436725.2 - 1 - 2 289 77
JO644608.2 - 1 - 2 294 77

View Full Product Details