Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCF20
Entrez Gene ID
102171958
Gene Name
transcription factor 20
Gene Aliases
-
Chromosome Location
Chr.5: 112076057 - 112171315 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048849.1 XP_017904338.1 4 - 5 6023 97

View Full Product Details