Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRG4
Entrez Gene ID
102184950
Gene Name
neuregulin 4
Gene Aliases
-
Chromosome Location
Chr.21: 30740287 - 30926333 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066097.1 XP_017921586.1 5 - 6 424 96

View Full Product Details