Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAFA4
Entrez Gene ID
102190363
Gene Name
TAFA chemokine like family member 4
Gene Aliases
FAM19A4
Chromosome Location
Chr.22: 32561361 - 32737028 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973472.2 XP_013828926.1 5 - 6 738 84

View Full Product Details