Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RFESD
Entrez Gene ID
102182973
Gene Name
Rieske Fe-S domain containing
Gene Aliases
-
Chromosome Location
Chr.7: 15559115 - 15595713 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050027.1 XP_017905516.1 2 - 3 331 96
GenBank mRNA JT760224.1 - 1 - 2 179 96

View Full Product Details