Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLN
Entrez Gene ID
102185321
Gene Name
motilin
Gene Aliases
-
Chromosome Location
Chr.23: 40761163 - 40766972 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696459.2 XP_005696516.1 3 - 4 345 59

View Full Product Details