Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11H2orf73
Entrez Gene ID
102185370
Gene Name
chromosome 11 C2orf73 homolog
Gene Aliases
-
Chromosome Location
Chr.11: 36653535 - 36675449 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013967722.2 XP_013823176.1 1 - 2 297 124

View Full Product Details