Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLB
Entrez Gene ID
102170862
Gene Name
klotho beta
Gene Aliases
-
Chromosome Location
Chr.6: 59118762 - 59150278 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681519.3 XP_005681576.2 1 - 2 847 72
GenBank mRNA JO426422.1 - 4 - 5 2348 72

View Full Product Details