Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPRC6A
Entrez Gene ID
102174303
Gene Name
G protein-coupled receptor class C group 6 member A
Gene Aliases
-
Chromosome Location
Chr.9: 21016467 - 21069713 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053511.1 XP_017909000.1 3 - 4 541 82

View Full Product Details